missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFYVE27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557987
This item is not returnable.
View return policy
Description
Immunogen sequence: RLRKRYPTNN FGNCTGCSAT FSVLKKRRSC SNCGNSFCSR CCSFKVPKSS MGATAPEAQR ETVFVCASCN QTLSK Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.
This gene encodes a protein with several transmembrane domains, a Rab11-binding domain and a lipid-binding FYVE finger domain. The encoded protein appears to promote neurite formation. A mutation in this gene has been reported to be associated with hereditary spastic paraplegia, however the pathogenicity of the mutation, which may simply represent a polymorphism, is unclear.
Specifications
| ZFYVE27 | |
| Polyclonal | |
| Unconjugated | |
| ZFYVE27 | |
| 2210011N02Rik; 9530077C24Rik; AI426636; AI593546; AI835681; im:7137347; LOW QUALITY PROTEIN: protrudin; PROTRUDIN; RP11-459F3.2; Spastic paraplegia 33 protein; SPG33; zfyve27; zgc:153156; zinc finger FYVE domain containing 27; zinc finger FYVE domain-containing protein 27; zinc finger FYVE-type containing 27; zinc finger, FYVE domain containing 27; zinc finger, FYVE domain containing 27 isoform c | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 118813 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T4F4 | |
| ZFYVE27 | |
| Recombinant protein corresponding to Human ZFYVE27. Recombinant protein control fragment (Product #RP-95663). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction