missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFYVE20/Rabenosyn 5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106714
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZFYVE20.
This gene encodes a zinc finger transcription factor. The encoded protein likely plays a role in transcriptional repression of interleukin 2. Mutations in this gene have been associated with posterior polymorphous corneal dystrophy-3 and late-onset Fuchs endothelial corneal dystrophy. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| ZFYVE20/Rabenosyn 5 | |
| Polyclonal | |
| Unconjugated | |
| Rbsn | |
| 110 kDa protein; 5330426D11Rik; FLJ34993; FYVE finger-containing Rab5 effector protein rabenosyn-5; MGC126210; RAB effector RBSN; Rabenosyn 5; rabenosyn, RAB effector; rabenosyn-5; Rbsn; ZFYVE20; zinc finger FYVE domain-containing protein 20; zinc finger, FYVE domain containing 20 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 312562, 64145, 78287 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q80Y56, Q9H1K0 | |
| Rbsn | |
| A synthesized peptide derived from human RBSN(Accession Q9H1K0), corresponding to amino acid residues R145-S195. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction