missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFYVE19 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23163-1-AP-150UL
This item is not returnable.
View return policy
Description
Key regulator of abscission step in cytokinesis: part of the cytokinesis checkpoint, a process required to delay abscission to prevent both premature resolution of intercellular chromosome bridges and accumulation of DNA damage. Together with CHMP4C, required to retain abscission-competent VPS4 (VPS4A and/or VPS4B) at the midbody ring until abscission checkpoint signaling is terminated at late cytokinesis. Deactivation of AURKB results in dephosphorylation of CHMP4C followed by its dissociation from ZFYVE19/ANCHR and VPS4 and subsequent abscissionSpecifications
| ZFYVE19 | |
| Polyclonal | |
| Unconjugated | |
| ZFYVE19 | |
| MPFYVE, ZFYVE19 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84936 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.65 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96K21 | |
| ZFYVE19 | |
| ZFYVE19 Fusion Protein Ag19593 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction