missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFYVE19 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | ZFYVE19 |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18263994
|
Novus Biologicals
NBP2-58993 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18614107
|
Novus Biologicals
NBP2-58993-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZFYVE19 Polyclonal specifically detects ZFYVE19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ZFYVE19 | |
| Unconjugated | |
| RUO | |
| FLJ14840, MLL partner containing FYVE domain, MPFYVE, zinc finger FYVE domain-containing protein 19, zinc finger, FYVE domain containing 19 | |
| ZFYVE19 | |
| IgG | |
| Affinity Purified |
| Polyclonal | |
| Rabbit | |
| Human | |
| 84936 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SGCLSFSAAVPRTGNTQQKVCKQCHEVLTRGSSANASKWSPPQNYKKRVAALEAKQKPSTSQSQGLTRQDQMIAERLARLRQENKPKLVPSQ | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title