missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFYVE16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13118-2-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes an endosomal protein that belongs to the FYVE zinc finger family of proteins. The encoded protein is thought to regulate membrane trafficking in the endosome. This protein functions as a scaffold protein in the transforming growth factor-beta signaling pathway and is involved in positive and negative feedback regulation of the bone morphogenetic protein signaling pathway. Alternate splicing results in multiple transcript variants.Specifications
| ZFYVE16 | |
| Polyclonal | |
| Unconjugated | |
| ZFYVE16 | |
| ENDOFIN, KIAA0305, ZFYVE16 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 218441, 499508, 9765 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7Z3T8, Q80U44 | |
| Zfyve16 | |
| ZFYVE16 Fusion Protein Ag3811 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering