missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFYVE16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557470
This item is not returnable.
View return policy
Description
Immunogen sequence: GAIGESHGIN IICETVDKQN TIENGLSLGE KSTIPVQQGL PTSKSEITNQ LSVSDINSQS VGGARPKQLF SLPSRTRSSK DLNKPDVPDT IESEPSTAD Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 54%.
The ZFYVE16 gene encodes endofin, an endosomal protein implicated in regulating membrane trafficking. It is characterized by the presence of a phosphatidylinositol 3-phosphate-binding FYVE domain positioned in the middle of the molecule.
Specifications
| ZFYVE16 | |
| Polyclonal | |
| Unconjugated | |
| ZFYVE16 | |
| AI035632; B130024H06Rik; B130031L15; Endofin; endosomal-associated FYVE domain protein; Endosome-associated FYVE domain protein; endosome-associated FYVE-domain protein; Kiaa0305; mKIAA0305; PPP1R69; protein phosphatase 1, regulatory subunit 69; RGD1564784; ZFYVE16; Zinc finger FYVE domain-containing protein 16; zinc finger FYVE-type containing 16; zinc finger, FYVE domain containing 16 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9765 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z3T8 | |
| ZFYVE16 | |
| Recombinant protein corresponding to Human ZFYVE16. Recombinant protein control fragment (Product #RP-96798). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction