missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFYVE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551768
This item is not returnable.
View return policy
Description
Immunogen sequence: IAPAYWRPNS QILSCNKCAT SFKDNDTKHH CRACGEGFCD SCSSKTRPVP ERGWGPAPVR VCDNCYEARN VQLAVTEAQV DDEGGTLIAR KVGEAVQNTL GAVVTAIDIP LGLVKDAARP AYWVPDHEIL HCHNCRKEFS IKLSKHH Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains two zinc-binding FYVE domains in tandem. This protein displays a predominantly Golgi, endoplasmic reticulum and vesicular distribution. Alternatively spliced transcript variants have been found for this gene, and they encode two isoforms with different sizes.
Specifications
| ZFYVE1 | |
| Polyclonal | |
| Unconjugated | |
| ZFYVE1 | |
| DFCP1; Double FYVE-containing protein 1; G630053K23; KIAA1589; mKIAA1589; phosphoinositide-binding protein SR3; PP10436; PPP1R172; protein phosphatase 1, regulatory subunit 172; SR3; TAFF1; Tandem FYVE fingers-1; tandem FYVE fingers-1 protein; Zfyve1; Zinc finger FYVE domain-containing protein 1; zinc finger FYVE-type containing 1; zinc finger protein, subfamily 2A, member 1; zinc finger, FYVE domain containing 1; ZNFN2A1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53349 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HBF4 | |
| ZFYVE1 | |
| Recombinant protein corresponding to Human ZFYVE1. Recombinant protein control fragment (Product #RP-89066). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction