missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFYVE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-84267-25ul
This item is not returnable.
View return policy
Description
ZFYVE1 Polyclonal antibody specifically detects ZFYVE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ZFYVE1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DFCP1, DFCP1phosphoinositide-binding protein SR3, Double FYVE-containing protein 1, KIAA1589tandem FYVE fingers-1 protein, SR3, TAFF1zinc finger FYVE domain-containing protein 1, Tandem FYVE fingers-1, zinc finger protein, subfamily 2A (FYVE domain containing), 1, zinc finger, FYVE domain containing 1, ZNFN2A1zinc finger protein, subfamily 2A, member 1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: IAPAYWRPNSQILSCNKCATSFKDNDTKHHCRACGEGFCDSCSSKTRPVPERGWGPAPVRVCDNCYEARNVQLAVTEAQVDDEGGTLIARKVGEAVQNTLGAVVTAIDIPLGLVKDAARPAYWVPDHEILHCHNCRKEFSIKLSKHH | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 53349 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction