missing translation for 'onlineSavingsMsg'
Learn More

ZFYVE1 Antibody (4B2), Novus Biologicals™

Product Code. 18391099 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391099 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18391099 Supplier Novus Biologicals Supplier No. H00053349M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZFYVE1 Monoclonal antibody specifically detects ZFYVE1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFYVE1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4B2
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_067083
Gene Alias DFCP1, DFCP1phosphoinositide-binding protein SR3, Double FYVE-containing protein 1, KIAA1589tandem FYVE fingers-1 protein, SR3, TAFF1zinc finger FYVE domain-containing protein 1, Tandem FYVE fingers-1, zinc finger protein, subfamily 2A (FYVE domain containing), 1, zinc finger, FYVE domain containing 1, ZNFN2A1zinc finger protein, subfamily 2A, member 1
Host Species Mouse
Immunogen ZFYVE1 (NP_067083.1, 1 a.a. ~ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSAQTSPAEKGLNPGLMCQESYACSGTDEAIFECDECCSLQCLRCEEELHRQERLRNHERIRLKPGHVPYCDLCKGLSGHLPGVRQRAIVRCQTCKINL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53349
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.