missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ZFYVE1 293T Cell Transient Overexpression Lysate

Product Code. 18386009 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386009 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386009 Supplier Novus Biologicals™ Supplier No. H00053349T03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Overexpression Lysate

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Target ZFYVE1
Synonym DFCP1, DFCP1phosphoinositide-binding protein SR3, Double FYVE-containing protein 1, KIAA1589tandem FYVE fingers-1 protein, SR3, TAFF1zinc finger FYVE domain-containing protein 1, Tandem FYVE fingers-1, zinc finger protein, subfamily 2A (FYVE domain containing), 1, zinc finger, FYVE domain containing 1, ZNFN2A1zinc finger protein, subfamily 2A, member 1
Gene Symbol ZFYVE1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.