missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Target | ZFYVE1 |
| Synonym | DFCP1, DFCP1phosphoinositide-binding protein SR3, Double FYVE-containing protein 1, KIAA1589tandem FYVE fingers-1 protein, SR3, TAFF1zinc finger FYVE domain-containing protein 1, Tandem FYVE fingers-1, zinc finger protein, subfamily 2A (FYVE domain containing), 1, zinc finger, FYVE domain containing 1, ZNFN2A1zinc finger protein, subfamily 2A, member 1 |
| Gene Symbol | ZFYVE1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?