missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ZFR |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
Description
ZFR Polyclonal specifically detects ZFR in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ZFR | |
| Unconjugated | |
| RUO | |
| FLJ41312, hZFR, zinc finger RNA binding protein | |
| ZFR | |
| IgG |
| Polyclonal | |
| Rabbit | |
| NP_035897 | |
| 51663 | |
| Synthetic peptide towards Zfr. Peptide sequence ICPVVSFTYVPSRLGEDAKMATGNYFGFTHSGAAAAAAAAQYSQQPASGV. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title