missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFPM1 Polyclonal Antibody

Description
Peptide sequence: SPAAAPEAVP PPPAPPSYSD KGVQTPSKGT PAPLPNGNHR YCRLCNIKFS Sequence homology: Cow: 79%; Human: 100%.
ZFPM1 is a transcription regulator that plays an essential role in erythroid and megakaryocytic cell differentiation. ZFPM1 is an essential cofactor that acts via the formation of a heterodimer with transcription factors of the GATA family GATA1, GATA2 and GATA3. Such heterodimer can both activate or repress transcriptional activity, depending on the cell and promoter context. The heterodimer formed with GATA proteins is essential to activate expression of genes such as NFE2, ITGA2B, alpha- and beta-globin, while it represses expression of KLF1. ZFPM1 may be involved in regulation of some genes in gonads. ZFPM1 may also be involved in cardiac development, in a non-redundant way with ZFPM2/FOG2.
Specifications
Specifications
| Antigen | ZFPM1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | ZFPM1 |
| Gene Accession No. | Q8IX07 |
| Gene Alias | FOG; Fog1; FOG-1; friend of GATA 1; friend of GATA protein 1; Friend of GATA-1; ZC2HC11A; ZFN89A; Zfpm1; Zinc finger protein 89A; Zinc finger protein multitype 1; Zinc finger protein ZFPM1; zinc finger protein, FOG family member 1; zinc finger protein, multitype 1; ZNF408; ZNF89A |
| Gene Symbols | ZFPM1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?