missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFPM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47572-25ul
This item is not returnable.
View return policy
Description
ZFPM1 Polyclonal specifically detects ZFPM1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ZFPM1 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| FOG-1, FOG1ZNF408, FOGfriend of GATA-1, Friend of GATA 1, Friend of GATA protein 1, ZFN89A, Zinc finger protein 89A, Zinc finger protein multitype 1, zinc finger protein ZFPM1, zinc finger protein, multitype 1, ZNF89A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ZFPM1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GFISTTRDILYSHLVTNHMVCQPGSKGEIYSPGAGHPATKLPPDSLGSFQQQHTALQGPLASADLGLAPTPSPGLDRKALAEATNGEARAAPQNGGSS | |
| 25 μL | |
| Vision | |
| 161882 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction