missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFP95 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575932
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. A member of the Krüppel C2H2-type zinc-finger protein family, ZKSCAN5, also known as ZFP95, is a 839 amino acid protein containing thirteen C2H2-type zinc fingers, one KRAB A domain and one SCAN box domain. ZKSCAN5 is located in the nucleus and is highly expressed in testis with lower levels of expression in liver, kidney, brain, striated muscle, lung and heart. Many KRAB A box-containing proteins have been shown to function as transcription repressors, suggesting a possible function of ZKSCAN5. As a result of alternative splicing, three transcripts of ZKSCAN5 exist, however one of those transcripts is specific to testis. The mouse homolog of ZKSCAN5 is differentially expressed in spermatogenesis.
Specifications
| ZFP95 | |
| Polyclonal | |
| Unconjugated | |
| ZKSCAN5 | |
| AI132486; AI326970; hKraba1; KIAA1015; ZFP95; ZFP-95; Zinc finger protein 95; zinc finger protein 95 homolog; zinc finger protein homologous to Zfp95 in mouse; zinc finger protein with KRAB and SCAN domains 5; zinc finger with KRAB and SCAN domains 5; Zkscan5; ZNF914; ZSCAN37 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22757, 23660, 304275 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2L8, Q9Z1D8 | |
| ZKSCAN5 | |
| Synthetic peptide corresponding to amino acids 284-329 of Human ZFP95. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction