missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP91 Mouse anti-Human, Clone:8C3D5, Abnova™
Mouse monoclonal antibody raised against recombinant human ZFP91.
Brand: Abnova MAB17247.100ug
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the zinc finger family of proteins. The gene product contains C2H2 type domains, which are the classical zinc finger domains found in numerous nucleic acid-binding proteins. A read-through transcript variant composed of ZFP91 and CNTF sequence has been identified, but it is thought to be non-coding. Read-through transcription of ZFP91 and CNTF has also been observed in mouse. A pseudogene has also been identified on chromosome 2. [provided by RefSeq]Specifications
| ZFP91 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunocytochemistry (1:100- 1:500) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| ZFP91 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 8C3D5 | |
| zinc finger protein 91 homolog (mouse) | |
| In PBS (0.05% sodium azide) | |
| FKSG11, PZF, ZNF757 | |
| Recombinant protein corresponding to amino acid 162-304 of human ZFP91 from E. coli. | |
| 80829 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction