missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFP91 Monoclonal Antibody (7G11H2)

Description
MA5-31957 has been tested in indirect ELISA.
ZFP91 is a member of the zinc finger family of proteins. The gene product contains C2H2 type domains, which are the classical zinc finger domains found in numerous nucleic acid-binding proteins. In addition to the monocistronic transcript originating from this locus, a co-transcribed variant composed of ZFP91 and CNTF sequence has been identified. The monocistronic and co-transcribed variants encode distinct isoforms. The co-transcription of ZFP91 and CNTF has also been observed in mouse.
Specifications
Specifications
| Antigen | ZFP91 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 7G11H2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | ZFP91 |
| Gene Accession No. | Q96JP5 |
| Gene Alias | 9130014I08Rik; A530054C17Rik; AL024263; AW545902; cocaine attenuated zinc-finger protein; DMS-8; drug resistance-associated sequence in melanoma 8; DSM-8; E3 ubiquitin-protein ligase ZFP91; FKSG11; Penta Zf protein; penta zinc finger protein; PZF; RING-type E3 ubiquitin transferase ZFP91; Zfp91; Zfp-91; ZFP91 zinc finger protein; ZFP91 zinc finger protein, atypical E3 ubiquitin ligase; zinc finger protein 757; zinc finger protein 91; zinc finger protein 91 homolog; zinc finger protein 91-like protein; zinc finger protein homologous to Zfp91 in mouse; Zinc finger protein PZF; ZNF757 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?