missing translation for 'onlineSavingsMsg'
Learn More

ZFP90 Antibody [DyLight 405], Novus Biologicals™

Product Code. 30733788 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733788 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733788 Supplier Novus Biologicals Supplier No. AF7870E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

ZFP90 Polyclonal antibody specifically detects ZFP90 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP90
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias KIAA1954, NK10, zfp-90, zinc finger protein 476, zinc finger protein 756, zinc finger protein 90 homolog, zinc finger protein 90 homolog (mouse), ZNF756
Host Species Sheep
Immunogen E. coli-derived recombinant human ZFP90, Lys94-Lys207, Accession # Q8TF47
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146198
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.