missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFP64 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568583
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 92%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 85%; Pig: 100%; Rabbit: 100%; Rat: 100%.
ZFP64 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers. ZFP64 may be involved in transcriptional regulation.
Specifications
| ZFP64 | |
| Polyclonal | |
| Unconjugated | |
| ZFP64 | |
| ZFP64; Zfp-64; ZFP64 zinc finger protein; zinc finger protein 338; Zinc finger protein 64; zinc finger protein 64 homolog; ZNF338 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55734 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NTW7 | |
| ZFP64 | |
| synthetic peptide directed towards the N terminal region of human ZFP64. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction