missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFP64 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530653
This item is not returnable.
View return policy
Description
Recommended positive controls: IMR32. Predicted reactivity: Mouse (96%), Rat (94%), Pig (98%), Rhesus Monkey (100%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ZFP64 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers. ZFP64 may be involved in transcriptional regulation.
Specifications
| ZFP64 | |
| Polyclonal | |
| Unconjugated | |
| ZFP64 | |
| ZFP64; Zfp-64; ZFP64 zinc finger protein; zinc finger protein 338; Zinc finger protein 64; zinc finger protein 64 homolog; ZNF338 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55734 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NTW7 | |
| ZFP64 | |
| Recombinant protein encompassing a sequence within the center region of human ZFP64. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction