missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Zfp57 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561456
This item is not returnable.
View return policy
Description
Immunogen sequence: PELITKLEQE EEQWREFVHL PNTEGLSEGK KKELREQHPS LRDEGTSDDK VFLACRGAGQ CPLSAPAGTM DRTRVLQASQ AGPPF Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 28%.
The protein encoded by this gene is a zinc finger protein containing a KRAB domain. Studies in mouse suggest that this protein may function as a transcriptional repressor. Mutations in this gene have been associated with transient neonatal diabetes mellitus type 1 (TNDM1).
Specifications
| Zfp57 | |
| Polyclonal | |
| Unconjugated | |
| ZFP57 | |
| bA145L22; bA145L22.2; C6orf40; DADB-25P22.4; G19; TNDM1; ZFP57; Zfp-57; ZFP57 zinc finger protein; zinc finger protein 57; Zinc finger protein 57 homolog; zinc finger protein 698; ZNF698 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 346171 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NU63 | |
| ZFP57 | |
| Recombinant protein corresponding to Human Zfp57. Recombinant protein control fragment (Product #RP-99612). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction