missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFP41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569730
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
ZFP41 gene ontology annotations related to this gene include nucleic acid binding.
Specifications
| ZFP41 | |
| Polyclonal | |
| Unconjugated | |
| ZFP41 | |
| CtFIN92; LOC100910508; uncharacterized LOC100910508; ZFP41; Zfp-41; ZFP41 zinc finger protein; zinc finger protein 41; Zinc finger protein 41 homolog; ZNF753 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 286128 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N8Y5 | |
| ZFP41 | |
| synthetic peptide directed towards the N-terminal region of human ZFP41. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction