missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP36L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-86905
This item is not returnable.
View return policy
Description
ZFP36L1 Polyclonal specifically detects ZFP36L1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ZFP36L1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 677 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ZBRK1KRAB zinc finger protein ZFQR, ZFQR, Zinc finger and BRCA1-interacting protein with a KRAB domain 1, zinc finger protein 350, Zinc finger protein ZBRK1, zinc-finger protein ZBRK1 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of human ZFP36L1 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction