missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP36 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12737-1-AP-150UL
This item is not returnable.
View return policy
Description
ZFP36 is a probable regulatory protein with a novel zinc finger structure involved in regulating the response to growth factors. Knockdown of ZFP36 increased both cognate macrophage gene mRNAs and inflammatory tumor necrosis factor protein release. Overexpression of ZFP36 resulted in decreased levels of the same genes supporting its role in regulating macrophage gene expression. Multiple phosphorylation sites in ZFP36 in mammalian cells were reported. ZFP36 can be phosphorylated by JNK as well as by the other members of the greater MAP kinase family. Gene expression profiling predicts clinical outcome of prostate cancer. Inappropriate ZFP36 production may be one factor that contributes to higher rheumatoid arthritis disease activity.Specifications
| ZFP36 | |
| Polyclonal | |
| Unconjugated | |
| ZFP36 | |
| G0S24, GOS24, NUP475, Protein TIS11A, RNF162A, TIS11, TIS11A, Tristetraproline, TTP, Zfp 36, ZFP36, Zinc finger protein 36 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22695, 7538, 79426 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22893, P26651, P47973 | |
| ZFP36 | |
| ZFP36 Fusion Protein Ag3461 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction