missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFP28 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569014
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 92%.
ZFP28 may be involved in transcriptional regulation. may have a role in embryonic development.
Specifications
| ZFP28 | |
| Polyclonal | |
| Unconjugated | |
| ZFP28 | |
| 2810438M17Rik; KIAA1431; Krueppel-like zinc finger factor X6; LOW QUALITY PROTEIN: zinc finger protein 28 homolog; mkr5; mkr-5; Protein mKR5; ZFP28; zfp-28; ZFP28 zinc finger protein; Zfp28l; zinc finger factor X6; zinc finger protein 28; Zinc finger protein 28 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140612 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NHY6 | |
| ZFP28 | |
| synthetic peptide directed towards the middle region of human ZFP28. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction