Learn More
Invitrogen™ ZFP28 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588435
Description
Immunogen sequence: MRGAASASVR EPTPLPGRGA PRTKPRAGRG PTVGTPATLA LPARGRPRSR NGLASKGQRG AAPTGPGHRA LPSRDTALPQ ERNKKLEAVG TGIEPKAMSQ GLVTFGDVAV DFSQEEWEWL NPIQRNLYRK VMLENYRNLA SLGLCVSKPD VISSLEQGKE PWTVKRKMTR AWCPDLKAVW KIKELPLKKD FCEGKLSQAV ITERLTSYNL EYSLLGEHWD YDALFETQPG LVTIKNLAVD FRQQLHPAQK NFCKNGIWEN NSDLGSAGHC VAKPDLVSLL EQEKEPWMVK RELTGSLFSG ECRRAWCGKS TARESGAFTK KAASSLIYSR KLYPGSLGLE RKLNSGSSK; Positive Samples: Mouse brain, Rat brain; Cellular Location: Nucleus.
ZFP28 may be involved in transcriptional regulation. may have a role in embryonic development.
Specifications
| ZFP28 | |
| Polyclonal | |
| Unconjugated | |
| ZFP28 | |
| 2810438M17Rik; KIAA1431; Krueppel-like zinc finger factor X6; LOW QUALITY PROTEIN: zinc finger protein 28 homolog; mkr5; mkr-5; Protein mKR5; ZFP28; zfp-28; ZFP28 zinc finger protein; Zfp28l; zinc finger factor X6; zinc finger protein 28; Zinc finger protein 28 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 102548693, 22690 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P10078 | |
| ZFP28 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-349 of human ZFP28 (NP_0012953691). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.