missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP112 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-37004
This item is not returnable.
View return policy
Description
ZFP112 Polyclonal specifically detects ZFP112 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications
| ZFP112 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| zfp-112, zinc finger protein 112 homolog, zinc finger protein 112 homolog (mouse), Zinc finger protein 228ZNF228 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7771 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence, ELISA | |
| 0.5 mg/mL | |
| Immunocytochemistry/Immunofluorescence 1:200, Peptide ELISA Detection limit 1:4000 | |
| NP_001076804.1 | |
| ZNF112 | |
| Peptide with sequence C-EREDDIENSHLKSYQ corresponding to internal region according to NP_001076804.1, NP_037512.3. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_001076804.1; NP_037512.3) | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction