missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFHX4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599757
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZFHX4.
May play a role in neural and muscle differentiation. May be involved in transcriptional regulation.
Specifications
| ZFHX4 | |
| Polyclonal | |
| Unconjugated | |
| ZFHX4 | |
| A930021B15; C130041O22Rik; LOW QUALITY PROTEIN: zinc finger homeobox protein 4; RGD1563022; ZFH4; Zfh-4; Zfhx4; ZHF4; zinc finger homeobox 4; zinc finger homeobox protein 4; zinc finger homeobox protein 4; LOW QUALITY PROTEIN: zinc finger homeobox protein 4; zinc finger homeodomain 4; zinc finger homeodomain protein 4; zinc-finger homeodomain protein 4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 310250, 79776, 80892 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86UP3, Q9JJN2 | |
| ZFHX4 | |
| A synthesized peptide derived from human ZFHX4(Accession Q86UP3), corresponding to amino acid residues D208-N258. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction