missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFHX3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118401
This item is not returnable.
View return policy
Description
ZFHX3 Polyclonal Antibody for Western Blot, Flow
Transcriptional repressor. It inhibits the enhancer element of the AFP gene by binding to its AT-rich core sequence. Regulator of myoblasts differentiation through the binding to the AT-rich sequence of MYF6 promoter and promoter repression. Down-regulates the MUC5AC promoter in gastric cancer. Miura Y., J. Biol. Chem. 270:26840-26848(1995). Morinaga T., Mol. Cell. Biol. 11:6041-6049(1991). Martin J., Nature 432:988-994(2004).
Specifications
| ZFHX3 | |
| Polyclonal | |
| Unconjugated | |
| ZFHX3 | |
| A230102L03Rik; alpha-fetoprotein enhancer binding protein; alpha-fetoprotein enhancer-binding protein; AT motif binding factor 1; AT motif-binding factor 1; ATBF1; AT-binding transcription factor 1; ATBT; C16orf47; mKIAA4228; Putative uncharacterized protein C16orf47; RGD1560268; WBP9; ZFH-3; Zfhx3; zinc finger homeobox 3; zinc finger homeobox protein 3; Zinc finger homeodomain protein 3; ZNF927 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 463 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q15911 | |
| ZFHX3 | |
| Synthetic peptide within human ATBF1 aa 2100-2200. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction