missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: SSNGRISPPA TSVSSLSESL PVQCTDGSVP EAQSALDSTS SSMQPSPVSN QSLLSESVAS SQLDSTSVDK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
ZA20D3 is a zinc-finger protein located on chromosome 15.
Specifications
Specifications
| Antigen | ZFAND6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.30 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZFAND6 |
| Gene Accession No. | Q6FIF0 |
| Gene Alias | 3110005P07Rik; AN1-type zinc finger protein 6; associated with PRK1 protein; associated with Prkcl1; Awp1; HT032; protein associated with PRK1; Za20d3; ZFAND5B; ZFAND6; zgc:101121; zinc finger A20 domain-containing protein 3; zinc finger AN1-type containing 6; zinc finger, A20 domain containing 3; zinc finger, AN1-type domain 6 |
| Gene Symbols | ZFAND6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
