missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFAND5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140210
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: CSTGCGFYGN PRTNGMCSVC YKEHLQR Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
ZNF216 (ZFAND5) is a potent inhibitory factor for osteoclast differentiation and the mechanism is unlikely due to direct attenuation of the NF-kappa B pathway. It also has redundant and distinct roles in regulating apoptosis.
Specifications
| ZFAND5 | |
| Polyclonal | |
| Unconjugated | |
| ZFAND5 | |
| 2310057A04Rik; 5830475F03Rik; AA415485; AN1-type zinc finger protein 5; EST00848 homolog; ubiquitin homolog; Za20d2; Zfand5; ZFAND5A; Zfp216; Zinc finger A20 domain-containing protein 2; zinc finger AN1-type containing 5; zinc finger protein 216; zinc finger, A20 domain containing 2; zinc finger, AN1-type domain 5; ZNF216 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7763 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O76080 | |
| ZFAND5 | |
| Recombinant protein corresponding to Human ZFAND5. Recombinant protein control fragment (Product #RP-109650). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction