missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFAND5 Antibody (OTI5H8), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74922CL1
This item is not returnable.
View return policy
Description
ZFAND5 Monoclonal antibody specifically detects ZFAND5 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| ZFAND5 | |
| Monoclonal | |
| CoraFluor 1 | |
| Zinc finger A20 domain-containing protein 2, Zinc finger protein 216ZA20D2, zinc finger, AN1-type domain 5, ZNF216A20 domain containing 2 | |
| Full length human recombinant protein of human ZFAND5 (NP_005998) produced in E.coli. | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 7763 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI5H8 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction