missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFAND3 Antibody (OTI1H10), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74921FR
This item is not returnable.
View return policy
Description
ZFAND3 Monoclonal specifically detects ZFAND3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ZFAND3 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| ZFAND3 | |
| Full length recombinant protein of human ZFAND3 (NP_068762) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1H10 | |
| Western Blot, Immunohistochemistry | |
| AN1-type zinc finger protein 3, FLJ17799, testis expressed sequence 27, Testis-expressed sequence 27, TEX27FLJ13222, zinc finger, AN1-type domain 3 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 60685.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction