missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Zeta Opioid Receptor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30451
This item is not returnable.
View return policy
Description
Zeta Opioid Receptor Polyclonal antibody specifically detects Zeta Opioid Receptor in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| Zeta Opioid Receptor | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| 7-60, 7-60 protein, OGFr, opioid growth factor receptor, Protein 7-60, Zeta-type opioid receptor | |
| Between 627 and 677 | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication, GPCR, Signal Transduction | |
| 11054 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction