missing translation for 'onlineSavingsMsg'
Learn More

Zeta Opioid Receptor Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£178.00 - £255.00

Specifications

Antigen Zeta Opioid Receptor
Dilution Immunocytochemistry/ Immunofluorescence 1:100-1:500
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18627092
View Documents
Novus Biologicals
NBP2-97488-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18671974
View Documents
Novus Biologicals
NBP2-97488-50ul
50 μg
£178.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zeta Opioid Receptor Polyclonal antibody specifically detects Zeta Opioid Receptor in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

Zeta Opioid Receptor
Immunofluorescence
Unconjugated
Rabbit
Cell Biology, Cell Cycle and Replication, GPCR, Signal Transduction
PBS (pH 7.0)
11054
IgG
Antigen and protein A Affinity-purified
Immunocytochemistry/ Immunofluorescence 1:100-1:500
Polyclonal
Purified
RUO
Human
7-60, 7-60 protein, OGFr, opioid growth factor receptor, Protein 7-60, Zeta-type opioid receptor
Produced in rabbits immunized with E. coli-derived Human Zeta Opioid Receptor fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.