missing translation for 'onlineSavingsMsg'
Learn More

TerraCycle™ Zero Waste Box™ Pipette Tip Boxes Recycling Solution, 5-pack Bundle, United Kingdom

Product Code. 32060650
Change view
Click to view available options
Capacity (Metric):
25 kg
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
32060650 25 kg Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32060650 Supplier TerraCycle™ Supplier No. CS180LUK_BUNDLE
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Support sustainability goals with this easy-to-use recycling solution for empty pipette tip boxes. Zero waste boxes are an all-inclusive eco-friendly waste disposal service which includes storage, shipping and processing of the materials.

  • Reduce the amount of waste sent to landfill or incineration.
  • Pack of 5 Large 140 L recyclable Zero Waste Boxes.
  • Empty pipette tip boxes, empty pipette tip racks.
  • DO NOT RECYCLE items that are contaminated with bodily fluids and materials considered hazardous or dangerous, autoclaved material, or medical waste.
  • Each box is supplied with a pre-affixed return shipping label.
  • How it works:
    • Order your 5-pack bundle
    • Collect: Fill the boxes with the accepted items.
    • Ship: Once full, use the pre-paid shipping label (already fastened to the boxes) to arrange for your waste to be collected for recycling.
    • Reorder: Order your next Zero Waste Boxes to continue to recycle.
  • Everything collected is sorted and processed into raw materials that can be reused instead of being sent to landfill or incineration.
  • Cardboard boxes are made from 100% recycled material.

Specifications

Capacity (Metric) 25 kg
Certifications/Compliance FSC
Capacity (English) 140 L
Color Brown, Green
Dimensions (L x W x H) 38 x 38 x 97 cm
Disposable Single-use
Height (Metric) 97 cm
Latex Free Yes
Length (Metric) 38 cm
Sterility Non-sterile
Width (Metric) 38 cm
Autoclavable Not Autoclavable
Includes Return Shipping Label
For Use With (Application) Pipette Tip Recycling Waste
Packaging United Kingdom
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less

An annual waste transfer note is required for waste originating from a commercial activity in the UK. Check guide on how to fill in the TerraCycle™ waste transfer note.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.