missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TerraCycle™ Laboratory PPE and Plastic Recycling Zero Waste Box™

Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
Description
Use this Zero Waste Box™ to recycle:
- Disposable PPE (face masks, gloves, garments, beard nets, disposable shoe covers, earplugs, hair nets)
- Protective eyewear (safety glasses)
- Rigid lab plastics
- Flexible lab plastics
- Disinfectant wipes
- We accept all types of disposable face masks made from plastic, such as dust masks, FFP2 (or so-called 'N95'), FFP3 and surgical masks.
- Do not send in:
- Medical waste. This includes material contaminated with blood, bodily fluids and waste, which should be collected in a hazardous clinical waste bin
- Lab plastics that are contaminated with hazardous waste
- Fabric, homemade or paper face masks
- Wax earplugs
- Plant-based and biogradable plastics and wipes
- Autoclaved material
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic)
- Batteries, pressurised canisters, broken glass
Specifications
Specifications
| Capacity (Metric) | 28 L |
| Capacity (English) | 1 cu.ft. |
| Color | Beige, Green, Red, White |
| Dimensions (L x W x H) | 25 x 25 x 46 cm |
| Disposable | Single-use |
| Latex Free | Yes |
| Size | Small |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Material | Cardboard, Plastic |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction