missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Zero Oxygen Solution Set

Product Code. 12784536
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 12784536

Brand: Hanna Instruments™ HI 7040

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Capacity (Metric) 500 mL
Type Hanna Instruments Zero Oxygen Solution Set
For Use With (Application) For calibrating both polarographic and galvanic dissolved oxygen sensors
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.