missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZER1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16647-1-AP-20UL
This item is not returnable.
View return policy
Description
ZER1 probably acts as target recruitment subunit in the E3 ubiquitin ligase complex ZER1-CUL2-Elongin BC.Specifications
| ZER1 | |
| Polyclonal | |
| Unconjugated | |
| ZER1 | |
| C9orf60, Hzyg, Protein zer 1 homolog, RP11 545E17.4, zer 1 homolog (C. elegans), ZER1, ZYG, Zyg 11 homolog B like protein, ZYG11BL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10444, 227693, 311842 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7Z7L7, Q80ZJ6 | |
| Zer1 | |
| ZER1 Fusion Protein Ag10031 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction