missing translation for 'onlineSavingsMsg'
Learn More

ZER1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£400.00

Specifications

Antigen ZER1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18263384
View Documents
Novus Biologicals
NBP1-69105
100 μL
£400.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZER1 Polyclonal specifically detects ZER1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZER1
Polyclonal
Rabbit
C9orf60, chromosome 9 open reading frame 60, homolog of Zyg-11, Hzygzyg-11 homolog B (C. elegans)-like, protein zer-1 homolog, zer-1 homolog (C. elegans), ZYG homolog, Zyg-11 homolog B-like protein, ZYG11BL, ZYGRP11-545E17.4
ZER1
IgG
88 kDa
Western Blot
Unconjugated
RUO
10444
Synthetic peptides corresponding to ZER1 (zer-1 homolog (C. elegans)) The peptide sequence was selected from the middle region of ZER1. Peptide sequence LTNSEYRSEQSVKLRRQVIQVVLNGMESYQEVTVQRNCCLTLCNFSIPEE.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.