missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zephirol-d7, TRC
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
Specifications
Specifications
| Chemical Name or Material | Zephirol-d7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C21H31D7ClN |
| Quantity | 100 mg |
| Synonym | N-Dodecyl-N,N-dimethylbenzenemethanaminium-d7 Chloride, Benzododecinium-d7 Chloride, Benzyldimethyldodecylammonium-d7 Chloride, Catigene OM-d7, Catinal CB 50-d7, Catiogen PAN-d7, Lauryldimethylbenzylammonium-d7 Chloride |
| InChI Formula | InChI=1S/C21H38N.ClH/c1-4-5-6-7-8-9-10-11-12-16-19-22(2,3)20-21-17-14-13-15-18-21;/h13-15,17-18H,4-12,16,19-20H2,1-3H3;1H/q+1;/p-1/i13D,14D,15D,17D,18D,20D2; |
| SMILES | [2H]C1=C(C([2H])([2H])[N+](C)(C)CCCCCCCCCCCC)C([2H])=C([2H])C([2H])=C1[2H].[Cl-] |
| Molecular Weight (g/mol) | 347.03 |
| Percent Purity | >95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction