missing translation for 'onlineSavingsMsg'
Learn More

Zeolite ZSM-5, ammonium

Product Code. 15452517
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452517 25 g 25g
15462517 100 g 100g
15472517 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15452517 Supplier Thermo Scientific Alfa Aesar Supplier No. 045880.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1318-02-1 | Al2O5Si | 162.043 g/mol

Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1318-02-1
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.043
MDL Number MFCD00132601
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
Synonym kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
PubChem CID 9942228
IUPAC Name dioxosilane;oxo(oxoalumanyloxy)alumane
SMILES O=[Al]O[Al]=O.O=[Si]=O

Specifications

Color White
Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Chemical Name or Material Zeolite ZSM-5, ammonium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.