missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zenocutuzumab Humanized Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Product Code. 17788793
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17788793 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17788793 Supplier Invitrogen™ Supplier No. MA542176

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level <0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Zenocutuzumab is under investigation in clinical trial NCT03321981 (MCLA-128 With Trastuzumab/chemotherapy in HER2+ and With Endocrine Therapy in ER+ and Low HER2 Breast Cancer).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zenocutuzumab Humanized
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 1.75 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias MCLA-128; PB4188; R040517
Host Species Human
Immunogen Human ERBB2/ERBB3.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.