Learn More
Carl Zeiss™ ZEN lite Module Time Lapse Hardware License Key
Description
- Requires Zen Lite (for modules) 410135-1001-330
- ZEN lite Module Time Lapse Module for acquisition of time series (time-lapse).
- Definition of intervals, total duration or number of cycles.
- Control via time or manually.
Specifications
Specifications
| Type | ZEN Lite Module Time Lapse Hardware License Key |
| For Use With (Equipment) | Acquisition of Time Series |
| Quantity | 1 Ea. |