Learn More
Paragon Scientific™ Zeitfuchs Cross-Arm Viscometer for Transparent and Opaque Liquids (UKAS ISO 17025 certified)
Click to view available options
Viscosity Range:
0.6 to 3 cSt
2 to 10 cSt
20 to 100 cSt
20,000 to 100,000 cSt
200 to 1000 cSt
2000 to 10,000 cSt
6 to 30 cSt
60 to 300 cSt
600 to 3000 cSt
6000 to 30,000 cSt
Unit Size:
Each
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
- Supplied with full UKAS ISO 17025 calibration
Specifications
Specifications
| Viscosity Range | 20 to 100 cSt |
| Calibrated | Yes |
| Approximate Constant | 0.1 |
| ASTM Numbers | D 445, D 446 |
| Certifications/Compliance | ASTM D 445, D 446 and ISO 3104, 3105 |
| Includes | UKAS ISO 17025 calibration |
| Size | 4 |
8 week lead-time