Learn More
Thermo Scientific™ Zeiss 2.5X Objective, Plan-Neofluar (0.075 NA)
Product Code. 17246583 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
Product Code. Quantity Unit Size
17246583 1 each Each
1 options
Product Code. 17246583 Supplier Thermo Scientific™ Supplier No. N010250
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
TRUSTED_SUSTAINABILITY
Specifications
Type Objective Lens
Product Line Zeiss
Description Zeiss 2.5X Objective, Plan-Neofluar (0.075 NA)
For Use With (Equipment) ArrayScan HCA Readers
Lens Type Dry
Magnification 2.5x (0.075NA)
Quantity 1 each