Learn More
Phenomenex™ Zebron Gas Management™ Wall-Mounting Clamp Set for Gas Traps
Product Code. 31743882 Shop All Phenomenex Products
Change view
Click to view available options
For Use With (Equipment):
Click-On Inline Filter
Unit Size:
Pack of 2
Product Code. For Use With (Equipment) Unit Size
31743882 Click-On Inline Filter Pack of 2
1 options
Product Code. 31743882 Supplier Phenomenex™ Supplier No. AG63180
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Zebron Gas Management™ filters and traps remove contaminants from mobile phase gases which reduces the potential for column bleed, increases the lifetime of your GC column, and protects sensitive GC detectors from damage.
  • Brand: Zebron Gas Management
  • Technique: Accessory
  • Phase: No Phase
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Click-On Inline Filter
Technique Gas Chromatography
Type Wall Mounting Clamp