Learn More
Phenomenex™ Zebron Gas Management™ O-Rings
Product Code. 31748605 Shop All Phenomenex Products
Change view
Click to view available options
Unit Size:
Pack of 20
Product Code. Unit Size
31748605 Pack of 20
1 options
Product Code. 31748605 Supplier Phenomenex™ Supplier No. AG63190
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Zebron Gas Management™ filters and traps remove contaminants from mobile phase gases which reduces the potential for column bleed, increases the lifetime of your GC column, and protects sensitive GC detectors from damage.
  • Brand: Zebron Gas Management
  • Product Type: O-Ring
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Click-On Inline Filter
Technique Gas Chromatography
Type O-rings