missing translation for 'onlineSavingsMsg'
Learn More

Phenomenex™ Zebron Gas Management™ High-Flow Connecting Unit for LC-MS (Hydrocarbon/Moisture)

Product Code. 31741636 Shop All Phenomenex Products
Change view
Click to view available options
For Use With (Equipment):
Click-On Inline Filter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31741636 Click-On Inline Filter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31741636 Supplier Phenomenex™ Supplier No. AG62204
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Zebron Gas Management™ filters and traps remove contaminants from mobile phase gases which reduces the potential for column bleed, increases the lifetime of your GC column, and protects sensitive GC detectors from damage.

  • Product Type: Connecting Unit
  • Format: 2-Position
  • For Use With: Gas Management Filters
  • Dectector Type: LC-MSD (Hydrocarbon/Moisture)
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Click-On Inline Filter
Technique Gas Chromatography
Type Double-ended Connector
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.