Learn More
Phenomenex™ Zebron Gas Management™ Click-On Inline Traps
Product Code. 31746763 Shop All Phenomenex Products
Change view
Click to view available options
Cartridge Type:
Combi (Moisture, Hydrocarbons)
Hydrocarbon
Moisture
Moisture, Oxygen, Hydrocarbon
Oxygen
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cartridge Type Unit Size
31746763 Moisture, Oxygen, Hydrocarbon Each
31744078 Oxygen Each
31745086 Moisture Each
31745630 Hydrocarbon Each
31741983 Combi (Moisture, Hydrocarbons) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31746763 Supplier Phenomenex™ Supplier No. AG63140
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Zebron Gas Management™ filters and traps remove contaminants from mobile phase gases which reduces the potential for column bleed, increases the lifetime of your GC column, and protects sensitive GC detectors from damage.
  • Attach a new filter in under 10 seconds
  • Optional electronic indicator display
  • Stainless steel to withstand high pressures
  • Secure wall-mounting using clamps
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type Moisture, Oxygen, Hydrocarbon
For Use With (Equipment) Click-On Inline Filter
Indicator Change Moisture: Yellow to Clear; Oxygen: Green to Gray
Technique Gas Chromatography
Type Indicating Triple Filter - Moisture, Oxygen, Hydrocarbons - He Preconditioned for GC/MS